Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical (R)-L-N-Fmoc-S-Mmt-alpha-Methylcysteine | 5 g | CAS 1198791-74-0 | InChIKey FTIUQZHUKYUVEE-LHEWISCISA-N

Catalog No. 502352649
Encompass

(R)-L-N-Fmoc-S-Mmt-alpha-Methylcysteine is a Amino Acid reagent (Subcategory: Alpha-alkylated) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1198791-74-0
- MDL: No data
- InChIKey: FTIUQZHUKYUVEE-LHEWISCISA-N
- Molecular Weight: 629.771
- Molecular Formula: C39H35NO5S
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29309095
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(((4-methoxyphenyl)diphenylmethyl)thio)-2-methylpropanoic acid
- SMILES: COC1=CC=C(C(C2=CC=CC=C2)(C3=CC=CC=C3)SC[C@](NC(OCC4C5=CC=CC=C5C6=CC=CC=C46)=O)(C(O)=O)C)C=C1

Catalog No. 50-235-2649 Supplier STA Pharmaceutical Supplier No. STAAA103705GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-L-N-Fmoc-S-Mmt-alpha-Methylcysteine is a Amino Acid reagent (Subcategory: Alpha-alkylated) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1198791-74-0
- MDL: No data
- InChIKey: FTIUQZHUKYUVEE-LHEWISCISA-N
- Molecular Weight: 629.771
- Molecular Formula: C39H35NO5S
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29309095
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(((4-methoxyphenyl)diphenylmethyl)thio)-2-methylpropanoic acid
- SMILES: COC1=CC=C(C(C2=CC=CC=C2)(C3=CC=CC=C3)SC[C@](NC(OCC4C5=CC=CC=C5C6=CC=CC=C46)=O)(C(O)=O)C)C=C1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.