Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-Lactamide 98.0+%, TCI America™
SDP

Catalog No. L01675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01675G 5 g
L016725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01675G Supplier TCI America Supplier No. L01675G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 598-81-2
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.094
MDL Number MFCD00210310
InChI Key SXQFCVDSOLSHOQ-UWTATZPHSA-N
Synonym (R)-(+)-2-Hydroxypropionamide
PubChem CID 1530236
IUPAC Name (2R)-2-hydroxypropanamide
SMILES CC(C(=O)N)O

Specifications

Melting Point 54°C
Quantity 5 g
Formula Weight 89.09
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-Lactamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.