Learn More
Medchemexpress LLC (R)-Lansoprazole-d4 (dexlansoprazole-d4) | 138530-94-6 | 99.13% | 373.39 | 1 MG
SDP
Catalog No. 5000348744
Encompass_Preferred
(R)-Lansoprazole-d4 is a deuterium-labeled variant of (R)-Lansoprazole, also known as Dexlansoprazole-d4. (R)-Lansoprazole is the R enantiomer of Lansoprazole, an orally active proton pump inhibitor that effectively reduces stomach acid production. Additionally, it acts as a potent brain-penetrant neutral sphingomyelinase (N-SMase) inhibitor, which functions as an exosome inhibitor. This compound is suitable for research purposes only.
  • Can be used as a tracer.
  • Can serve as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Features stable heavy isotopes incorporated into drug molecules.
  • Deuteration may influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-48744 Supplier Medchemexpress LLC Supplier No. HY13662BS1MG
Add to Cart
Add to Cart
(R)-Lansoprazole-d4 is a deuterium-labeled variant of (R)-Lansoprazole, also known as Dexlansoprazole-d4. (R)-Lansoprazole is the R enantiomer of Lansoprazole, an orally active proton pump inhibitor that effectively reduces stomach acid production. Additionally, it acts as a potent brain-penetrant neutral sphingomyelinase (N-SMase) inhibitor, which functions as an exosome inhibitor. This compound is suitable for research purposes only.
  • Can be used as a tracer.
  • Can serve as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Features stable heavy isotopes incorporated into drug molecules.
  • Deuteration may influence pharmacokinetic and metabolic profiles of drugs.