Learn More
Medchemexpress LLC (R)-Lercanidipine-d3 hydrochloride | 1217724-52-1 | 99.9% | 5 MG
SDP
Catalog No. 5000408317
Encompass_Preferred
(R)-lercanidipine D3 hydrochloride is a deuterium labeled (R)-Lercanidipine hydrochloride. (R)-Lercanidipine-d3 (hydrochloride), the R-enantiomer of Lercanidipine, is a calcium channel blocker. It is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08317 Supplier Medchemexpress LLC Supplier No. HYB0612DS5MG
(R)-lercanidipine D3 hydrochloride is a deuterium labeled (R)-Lercanidipine hydrochloride. (R)-Lercanidipine-d3 (hydrochloride), the R-enantiomer of Lercanidipine, is a calcium channel blocker. It is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.