Learn More
Medchemexpress LLC (R)-Leucic acid | 20312-37-2 | 97% | 132.16 | 500 MG
SDP
Catalog No. 5000404797
Encompass_Preferred
(R)-Leucic acid (Standard) is an analytical standard for research and analytical applications. It is the D-isomer of the α-hydroxy analogue of Leucine, a metabolite of Lactobacillus that can promote intestinal fatty acid absorption. This compound is used to study microbe-host interactions and the regulation of lipid metabolism by probiotics.
  • Intended for research and analytical applications
  • Promotes intestinal fatty acid absorption by upregulating CD36 expression
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Suitable for studying microbe-host interactions and lipid metabolism regulation by probiotics
Catalog No. 50-004-04797 Supplier Medchemexpress LLC Supplier No. HY30216500MG
Add to Cart
Add to Cart
(R)-Leucic acid (Standard) is an analytical standard for research and analytical applications. It is the D-isomer of the α-hydroxy analogue of Leucine, a metabolite of Lactobacillus that can promote intestinal fatty acid absorption. This compound is used to study microbe-host interactions and the regulation of lipid metabolism by probiotics.
  • Intended for research and analytical applications
  • Promotes intestinal fatty acid absorption by upregulating CD36 expression
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Suitable for studying microbe-host interactions and lipid metabolism regulation by probiotics