Learn More
Medchemexpress LLC (R)-Leucic acid (D-α-Hydroxyisocaproic acid) | 20312-37-2 | 98.0% | 132.16 | 100 G
SDP
Catalog No. 5000404446
Encompass_Preferred
(R)-Leucic acid, also known as D-α-Hydroxyisocaproic acid, is an orally active D-isomer that functions as an α-hydroxy analogue of Leucine. This compound is a metabolite produced by Lactobacillus and has been shown to enhance intestinal fatty acid absorption through the upregulation of CD36 expression. It serves as a valuable tool for researchers investigating microbe-host interactions and the mechanisms by which probiotics influence lipid metabolism.
  • Orally active D-isomer of Leucine's α-hydroxy analogue.
  • Functions as a metabolite of Lactobacillus.
  • Promotes intestinal fatty acid absorption via CD36 upregulation.
  • Useful for studying microbe-host interactions.
  • Applicable in research concerning the regulation of lipid metabolism by probiotics.
Catalog No. 50-004-04446 Supplier Medchemexpress LLC Supplier No. HY30216100G
(R)-Leucic acid, also known as D-α-Hydroxyisocaproic acid, is an orally active D-isomer that functions as an α-hydroxy analogue of Leucine. This compound is a metabolite produced by Lactobacillus and has been shown to enhance intestinal fatty acid absorption through the upregulation of CD36 expression. It serves as a valuable tool for researchers investigating microbe-host interactions and the mechanisms by which probiotics influence lipid metabolism.
  • Orally active D-isomer of Leucine's α-hydroxy analogue.
  • Functions as a metabolite of Lactobacillus.
  • Promotes intestinal fatty acid absorption via CD36 upregulation.
  • Useful for studying microbe-host interactions.
  • Applicable in research concerning the regulation of lipid metabolism by probiotics.