Learn More
Sigma Aldrich Fine Chemicals Biosciences (R)-(+)-Limonene analytical standard | 5989-27-5 | MFCD00062991 | 1ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 621181ML
Encompass
(R)-(+)-Limonene analytical standard | Purity: >=99.0% (sum of enantiomers, GC) | Mol Wt: 136.23 | 5989-27-5 | MFCD00062991 | 1ML
Catalog No. NC1048234