Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC R-(+)-α-lipoic acid | 1200-22-2 | MFCD01631142 | 99.4% | 206.33 | C8H14O2S2 | 10 MG
SDP

Catalog No. 5000294715
Encompass_Preferred

Analytical standard of (R)-(+)-α-lipoic acid intended for research and analytical applications such as HPLC, GC, and MS. It is used as a quantitative and qualitative reference material for method development, calibration, and validation in biochemical and pharmaceutical analysis.

  • High purity suitable for analytical assays (99.36%).
  • Defined chemical identity for accurate reference (C8H14O2S2, MW 206.33).
  • Compact 10 mg packaging for analytical use and method validation.
  • Stable under recommended storage; consult the certificate of analysis for handling.

Catalog No. 50-002-94715 Supplier Medchemexpress LLC Supplier No. HY18733R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical standard of (R)-(+)-α-lipoic acid intended for research and analytical applications such as HPLC, GC, and MS. It is used as a quantitative and qualitative reference material for method development, calibration, and validation in biochemical and pharmaceutical analysis.

  • High purity suitable for analytical assays (99.36%).
  • Defined chemical identity for accurate reference (C8H14O2S2, MW 206.33).
  • Compact 10 mg packaging for analytical use and method validation.
  • Stable under recommended storage; consult the certificate of analysis for handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.