Learn More
Cayman Chemical (R)-Lansoprazole, Purity: ≥98%, CAS: 138530-94-6, 50 mg
SDP
Catalog No. 502966700
Encompass_Preferred
A pure enantiomer of lansoprazole that inhibits acid formation in isolated canine parietal cells with an IC50 value of 59 nM and inhibits the H+/K+-ATPase with an IC50 value of 4.2 µM
Catalog No. 50-296-6700 Supplier Cayman Chemical Supplier No. 1823550 MG
A pure enantiomer of lansoprazole that inhibits acid formation in isolated canine parietal cells with an IC50 value of 59 nM and inhibits the H+/K+-ATPase with an IC50 value of 4.2 µM