Learn More
Cayman Chemical R-MephenytoIn 10mg
SDP
Catalog No. 502933057
Encompass_Preferred
The (R) enantiomer of mephenytoin; can be demethylated by CYP2C9 to form the active metabolite 5-ethyl-5-phenylhydantoin (nirvanol); the ratio of (S)- to (R)-mephenytoin in urine following administration of the racemic mixture has been used to detect polymorphisms in drug metabolism by CYP2C19, as only (S)-mephenytoin is a substrate of CYP2C19
Catalog No. 50-293-3057 Supplier Cayman Chemical Supplier No. 2589110 MG
The (R) enantiomer of mephenytoin; can be demethylated by CYP2C9 to form the active metabolite 5-ethyl-5-phenylhydantoin (nirvanol); the ratio of (S)- to (R)-mephenytoin in urine following administration of the racemic mixture has been used to detect polymorphisms in drug metabolism by CYP2C19, as only (S)-mephenytoin is a substrate of CYP2C19