Learn More
Medchemexpress LLC R-mephenytoin | 71140-51-7 | MFCD00022407 | 99.6% | 218.25 g·mol-1 | C12H14N2O2 | 50MG

Supplier: Medchemexpress LLC HY12604350MG
(R)-Mephenytoin is the R-enantiomer of mephenytoin, a hydantoin-class anticonvulsant used as a research reagent. The compound is intended for laboratory research use only and is not for clinical or human use.
- R-enantiomer of mephenytoin
- High reported purity: 99.6%
- Chemical formula C12H14N2O2
- Molecular weight 218.25 g·mol-1
- Available in small-mass formats suitable for analytical and pharmacology studies
- Supplied as a solid research chemical
- For research use only; not for human consumption
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.