Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-mephenytoin | 71140-51-7 | MFCD00272651 | 99.6% | 218.25 g/mol | C12H14N2O2 | 25MG
SDP

Supplier:  Medchemexpress LLC HY12604325MG

Encompass_Preferred

(R)-Mephenytoin is the R-enantiomer of the anticonvulsant mephenytoin, provided as a high-purity research reagent for pharmacology, metabolic, and analytical studies. It is typically used as a reference standard or investigational compound in studies of drug metabolism (CYP-mediated demethylation) and anticonvulsant pharmacology; it is not intended for human use.

  • R-enantiomer of mephenytoin.
  • High purity: 99.6%.
  • Molecular formula: C12H14N2O2.
  • Molecular weight: 218.25 g/mol.
  • Available in multiple sizes and as a DMSO solution for convenience.
  • Commonly used as a reference standard in metabolic and CYP enzyme studies.
  • Intended for research use only; not for clinical or human use.

Catalog No. 50-002-35032


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.