Learn More
eMolecules​ 39637-99-5 | (R)-3,3,3-TRIFLUORO-2-METHOXY-2-PHENYLPROPANOYL CHLORIDE (CA. 18% IN DICHLOROMETHANE, CA. 1.0MOL/L) [FOR DETERMINATION OF THE OPTICAL PURITY OF ALCOHOLS AND AMINES] | AstaTech | MFCD00044400 | 252.620 | C10H8ClF3O2 | 98.000 | CO[C@](C(Cl)=O)(c1ccccc1)C(F)(F)F | 0.1g | 721428415
Catalog No. 502457650 Shop All eMolecules Inc. Products
Encompass_Preferred
(R)-3,3,3-TRIFLUORO-2-METHOXY-2-PHENYLPROPANOYL CHLORIDE (CA. 18% IN DICHLOROMETHANE, CA. 1.0MOL/L) [FOR DETERMINATION OF THE OPTICAL PURITY OF ALCOHOLS AND AMINES] | AstaTech | 39637-99-5 | MFCD00044400 | 252.620 | C10H8ClF3O2 | 98.000 | CO[C@](C(Cl)=O)(c1ccccc1)C(F)(F)F | 0.1g | 721428415
Catalog No. 50-245-7650 Supplier eMolecules​ Supplier No. 721428415
(R)-3,3,3-TRIFLUORO-2-METHOXY-2-PHENYLPROPANOYL CHLORIDE (CA. 18% IN DICHLOROMETHANE, CA. 1.0MOL/L) [FOR DETERMINATION OF THE OPTICAL PURITY OF ALCOHOLS AND AMINES] | AstaTech | 39637-99-5 | MFCD00044400 | 252.620 | C10H8ClF3O2 | 98.000 | CO[C@](C(Cl)=O)(c1ccccc1)C(F)(F)F | 0.1g | 721428415