Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-methyl 2-amino-4-methylpentanoate hydrochloride | 5845-53-4 | MFCD00066115 | ≥98.0% | 181.66 g·mol⁻¹ | C7H16ClNO2 | 100 G
SDP

Catalog No. 5000302080
Encompass_Preferred

(R)-methyl 2-amino-4-methylpentanoate hydrochloride is the R-enantiomer methyl ester hydrochloride of leucine supplied as a solid research reagent. It is used as a chiral building block in organic synthesis, peptide derivatization, and medicinal chemistry research, and can serve as a reference or intermediate in analytical and synthetic workflows.

  • R-enantiomer methyl ester hydrochloride suitable for chiral synthesis.
  • Purity ≥98.0%.
  • Molecular weight 181.66 g·mol⁻¹ and formula C7H16ClNO2.
  • Solid form for convenient handling and storage.
  • Useful as a building block for peptide and medicinal chemistry applications.

Catalog No. 50-003-02080 Supplier Medchemexpress LLC Supplier No. HYW016425100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-methyl 2-amino-4-methylpentanoate hydrochloride is the R-enantiomer methyl ester hydrochloride of leucine supplied as a solid research reagent. It is used as a chiral building block in organic synthesis, peptide derivatization, and medicinal chemistry research, and can serve as a reference or intermediate in analytical and synthetic workflows.

  • R-enantiomer methyl ester hydrochloride suitable for chiral synthesis.
  • Purity ≥98.0%.
  • Molecular weight 181.66 g·mol⁻¹ and formula C7H16ClNO2.
  • Solid form for convenient handling and storage.
  • Useful as a building block for peptide and medicinal chemistry applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.