Learn More
Medchemexpress LLC HY-13259C 5mg Medchemexpress, (R)-MG-132 CAS:1211877-36-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13259C5MG
Encompass_Preferred
Medchemexpress, HY-13259C 5mg (R)-MG-132 CAS:1211877-36-9 (R)-MG-132 is the enantiomer of MG-132. (R)-MG-132 is a proteasome inhibitor with weaker cell cytotoxicity than MG-132 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2657