Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC R-mirtazapine | 61364-37-2 | 99.8% | 265.35 g·mol⁻¹ | C17H19N3 | 10 MG
SDP

Catalog No. 5000297301
Encompass_Preferred

(R)-Mirtazapine is the R-enantiomer of mirtazapine, described as a 5-HT3 receptor antagonist with reported antinociceptive activity. It is supplied as a high-purity solid research reagent intended for laboratory use.

  • High purity: 99.8%.
  • Chiral R-enantiomer with 5-HT3 receptor antagonist activity.
  • Chemical formula C17H19N3; molecular weight 265.35 g·mol⁻¹.
  • CAS number 61364-37-2 for substance identification.
  • Supplied as a solid powder suitable for analytical and preclinical research.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months).

Catalog No. 50-002-97301 Supplier Medchemexpress LLC Supplier No. HYB0352B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Mirtazapine is the R-enantiomer of mirtazapine, described as a 5-HT3 receptor antagonist with reported antinociceptive activity. It is supplied as a high-purity solid research reagent intended for laboratory use.

  • High purity: 99.8%.
  • Chiral R-enantiomer with 5-HT3 receptor antagonist activity.
  • Chemical formula C17H19N3; molecular weight 265.35 g·mol⁻¹.
  • CAS number 61364-37-2 for substance identification.
  • Supplied as a solid powder suitable for analytical and preclinical research.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.