Learn More
Medchemexpress LLC (R)-Mirtazapine ((R)-Org3770) | 61364-37-2 | 99.8%
SDP
Catalog No. 5000406609
Encompass_Preferred
(R)-Mirtazapine is an R(+)-enantiomer of Mirtazapine with antinociceptive properties. It functions as a 5-HT3 receptor antagonist and is primarily metabolized by CYP3A4.
  • Acts as a 5-HT3 receptor antagonist
  • Primarily metabolized by CYP3A4
  • Exhibits antinociceptive properties
  • Suitable for research use only
Catalog No. 50-004-06609 Supplier Medchemexpress LLC Supplier No. HYB0352B5MG
(R)-Mirtazapine is an R(+)-enantiomer of Mirtazapine with antinociceptive properties. It functions as a 5-HT3 receptor antagonist and is primarily metabolized by CYP3A4.
  • Acts as a 5-HT3 receptor antagonist
  • Primarily metabolized by CYP3A4
  • Exhibits antinociceptive properties
  • Suitable for research use only