Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse IL-6R alpha Quantikine ELISA Kit
SDP

Catalog No. MR600
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MR600 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MR600 Supplier R&D Systems™ Supplier No. MR600

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

a 4.5 hour solid phase ELISA designed to measure mouse IL-6 R alpha levels in cell culture supernates, serum, and heparin plasma.

The Mouse IL-6R alpha Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse IL-6R alpha in cell culture supernates (50 ul), serum (10 ul), heparin plasma (10 ul).

Specifications

Assay Range 0.19 - 12ng/mL
Assay Sensitivity 0.07ng/mL
Detectable Analytes IL-6 R alpha
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Strip Plate
Sample Type Cell Culture Supernatant, Plasma (Heparin), Serum
Specificity Natural and Recombinant Mouse IL-6 R alpha
For Use With (Application) ELISA
Gene ID (Entrez) 16194
Host Species Mouse
Measuring Range 0.188-12ng/mL
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6 R-1, IL6RA, IL-6 RA, IL6RQ, interleukin 6 receptor
Target IL-6R alpha
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.