Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse/Rat cAMP Parameter Assay Kit

Catalog No. kge012b
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KGE012B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. KGE012B Supplier R&D Systems™ Supplier No. KGE012B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse/Rat cAMP Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse/Rat cAMP Parameter Assay Kit quantifies mouse, rat cAMP in cell culture supernates (50 ul), cell lysates (50 ul), tissue homogenates (50 ul), edta plasma (50 ul), urine (10 ul).

Specifications

Assay Range 3.75 - 240 nmol/L
Assay Sensitivity 1.54 nmol/L
Detectable Analytes cAMP
Assay Solid Phase Competitive ELISA
Conjugate Biotin
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Strip Plate
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA), Tissue Homogenates, Urine
Specificity Cyclic AMP in Mouse and Rat Samples
For Use With (Application) ELISA
Host Species Mouse, Rat
Measuring Range 3.75-240 nmol/L
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (50uL),Tissue Homogenates (50uL),EDTA Plasma (50uL),Urine (10uL)
Storage Requirements Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.