Learn More
Medchemexpress LLC (R)-N-(1-(3-methoxyphenyl)ethyl)-4-(pyridin-4-yl)benzamide | 1127308-52-4 | 99.9% | 50 MG
SDP
Catalog No. 5000234261
Encompass_Preferred
ROCK inhibitor-2 is a selective dual inhibitor of Rho-associated kinases ROCK1 and ROCK2 used for laboratory research. It inhibits ROCK-mediated phosphorylation such as pMYPT1 and has reported antitumor activity in preclinical studies. The compound is supplied as a solid or as a DMSO solution and is intended for research use only.
  • Selective dual inhibitor of ROCK1 and ROCK2.
  • Inhibits phosphorylation of pMYPT1.
  • High purity (99.9%).
  • Molecular weight 332.40 g·mol⁻¹, formula C21H20N2O2.
  • Available as solid or 10 mM solution in DMSO.
  • Intended for laboratory research use only.
Catalog No. 50-002-34261 Supplier Medchemexpress LLC Supplier No. HY11993750MG
ROCK inhibitor-2 is a selective dual inhibitor of Rho-associated kinases ROCK1 and ROCK2 used for laboratory research. It inhibits ROCK-mediated phosphorylation such as pMYPT1 and has reported antitumor activity in preclinical studies. The compound is supplied as a solid or as a DMSO solution and is intended for research use only.
  • Selective dual inhibitor of ROCK1 and ROCK2.
  • Inhibits phosphorylation of pMYPT1.
  • High purity (99.9%).
  • Molecular weight 332.40 g·mol⁻¹, formula C21H20N2O2.
  • Available as solid or 10 mM solution in DMSO.
  • Intended for laboratory research use only.