Learn More
Medchemexpress LLC (R)-N-(1-(4-fluorophenyl)ethyl)-3-(2-(isoquinolin-3-yl)-1H-imidazol-5-yl)benzamide | 950225-08-8 | 99.9% | 436.48 | C27H21FN4O | 10MG
SDP
Catalog No. 5000236488
Encompass_Preferred
TTP-8307 is a small-molecule antiviral research reagent that inhibits replication of several rhino- and enteroviruses by targeting the oxysterol-binding protein (OSBP) and interfering with viral RNA synthesis. It demonstrates activity against coxsackievirus B3 and poliovirus and is supplied as a solid, high-purity compound for laboratory use.
  • Potent inhibitor of rhino- and enterovirus replication.
  • Acts via oxysterol-binding protein (OSBP) and disrupts viral RNA synthesis.
  • Reported activity against coxsackievirus B3 and poliovirus.
  • High purity suitable for research applications (≈99.9%).
  • Supplied as a solid powder with recommended cold storage.
  • Available in small pack sizes for laboratory experiments (for example, 10 mg).
Catalog No. 50-002-36488 Supplier Medchemexpress LLC Supplier No. HY12480610MG
TTP-8307 is a small-molecule antiviral research reagent that inhibits replication of several rhino- and enteroviruses by targeting the oxysterol-binding protein (OSBP) and interfering with viral RNA synthesis. It demonstrates activity against coxsackievirus B3 and poliovirus and is supplied as a solid, high-purity compound for laboratory use.
  • Potent inhibitor of rhino- and enterovirus replication.
  • Acts via oxysterol-binding protein (OSBP) and disrupts viral RNA synthesis.
  • Reported activity against coxsackievirus B3 and poliovirus.
  • High purity suitable for research applications (≈99.9%).
  • Supplied as a solid powder with recommended cold storage.
  • Available in small pack sizes for laboratory experiments (for example, 10 mg).