Learn More
Medchemexpress LLC (R)-N-(1-(4-fluorophenyl)ethyl)-3-(2-(isoquinolin-3-yl)-1H-imidazol-5-yl)benzamide | 950225-08-8 | 99.9% | 436.48 | C27H21FN4O | 50MG
SDP
Catalog No. 5000235853
Encompass_Preferred
TTP-8307 is a small-molecule research antiviral that inhibits replication of several rhino- and enteroviruses by targeting oxysterol-binding protein (OSBP). It shows activity against coxsackievirus B3 (EC50 = 1.2 μM) and poliovirus and is supplied as a high-purity reagent for in vitro studies.
  • Potent antiviral activity via oxysterol-binding protein inhibition.
  • High purity (99.9%).
  • Molecular formula C27H21FN4O; molecular weight 436.48 g/mol.
  • Available in multiple package sizes including 50 mg.
  • Powder stable at -20°C for long-term storage; different stability in solvent.
Catalog No. 50-002-35853 Supplier Medchemexpress LLC Supplier No. HY12480650MG
TTP-8307 is a small-molecule research antiviral that inhibits replication of several rhino- and enteroviruses by targeting oxysterol-binding protein (OSBP). It shows activity against coxsackievirus B3 (EC50 = 1.2 μM) and poliovirus and is supplied as a high-purity reagent for in vitro studies.
  • Potent antiviral activity via oxysterol-binding protein inhibition.
  • High purity (99.9%).
  • Molecular formula C27H21FN4O; molecular weight 436.48 g/mol.
  • Available in multiple package sizes including 50 mg.
  • Powder stable at -20°C for long-term storage; different stability in solvent.