Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-N-(1-(Naphthalen-1-yl)ethyl)-3-(3-(trifluoromethyl)phenyl)propanamide | 1005450-55-4 | 99.05% | C22H20F3NO | 5 G
SDP

Catalog No. 5000433866
Encompass_Preferred

This compound is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only and not sold to patients.

  • Appearance: Solid, white to off-white
  • Purity: 99.05%
  • Storage conditions: 4°C, sealed storage, away from moisture

Catalog No. 50-004-33866 Supplier Medchemexpress LLC Supplier No. HY753895G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This compound is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only and not sold to patients.

  • Appearance: Solid, white to off-white
  • Purity: 99.05%
  • Storage conditions: 4°C, sealed storage, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.