Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-N-(1-(naphthalen-1-yl)ethyl)-3-(3-(trifluoromethyl)phenyl)propanamide | 1005450-55-4 | 99.1% | C22H20F3NO | 10 G
SDP

Catalog No. 5000434313
Encompass_Preferred

(R)-N-(1-(Naphthalen-1-yl)ethyl)-3-(3-(trifluoromethyl)phenyl)propanamide is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.

  • Solid appearance, white to off-white
  • Molecular weight: 371.40
  • Purity: 99.05%
  • Soluble in DMSO (16.67 mg/mL)
  • Used as a drug intermediate

Catalog No. 50-004-34313 Supplier Medchemexpress LLC Supplier No. HY7538910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-N-(1-(Naphthalen-1-yl)ethyl)-3-(3-(trifluoromethyl)phenyl)propanamide is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.

  • Solid appearance, white to off-white
  • Molecular weight: 371.40
  • Purity: 99.05%
  • Soluble in DMSO (16.67 mg/mL)
  • Used as a drug intermediate

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.