Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical (R)-N-Fmoc-2-amino-oct-7-ynoic acid | 10 g | CAS 2350405-99-9 | InChIKey JGAAQTGBSLHOGR-OAQYLSRUSA-N

Catalog No. 502349680
Encompass

(R)-N-Fmoc-2-amino-oct-7-ynoic acid is a Amino Acid reagent (Subcategory: Azido/Alkynyl AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 2350405-99-9
- MDL: No data
- InChIKey: JGAAQTGBSLHOGR-OAQYLSRUSA-N
- Molecular Weight: 377.44
- Molecular Formula: C23H23NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)oct-7-ynoic acid
- SMILES: C#CCCCC[C@@H](NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)C(O)=O

Catalog No. 50-234-9680 Supplier STA Pharmaceutical Supplier No. STAAA1029910GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-N-Fmoc-2-amino-oct-7-ynoic acid is a Amino Acid reagent (Subcategory: Azido/Alkynyl AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 2350405-99-9
- MDL: No data
- InChIKey: JGAAQTGBSLHOGR-OAQYLSRUSA-N
- Molecular Weight: 377.44
- Molecular Formula: C23H23NO4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)oct-7-ynoic acid
- SMILES: C#CCCCC[C@@H](NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)C(O)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.