Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical (R)-N-Fmoc-3-amino-4-(phenylthio)butanoic Acid | 50 g | CAS 1244724-97-7 | MDL MFCD18070978

Catalog No. 502350415
Encompass

(R)-N-Fmoc-3-amino-4-(phenylthio)butanoic Acid is a Amino Acid reagent (Subcategory: Beta AA) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1244724-97-7
- MDL: MFCD18070978
- InChIKey: ZXVKPJJQNJPHQT-QGZVFWFLSA-N
- Molecular Weight: 433.522
- Molecular Formula: C25H23NO4S
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29309095
- Country Of Origin: China
- IUPAC: (R)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-(phenylthio)butanoic acid
- SMILES: O=C(C[C@H](CSC1=CC=CC=C1)NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)O

Catalog No. 50-235-0415 Supplier STA Pharmaceutical Supplier No. STAAA1028350GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-N-Fmoc-3-amino-4-(phenylthio)butanoic Acid is a Amino Acid reagent (Subcategory: Beta AA) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1244724-97-7
- MDL: MFCD18070978
- InChIKey: ZXVKPJJQNJPHQT-QGZVFWFLSA-N
- Molecular Weight: 433.522
- Molecular Formula: C25H23NO4S
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29309095
- Country Of Origin: China
- IUPAC: (R)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-(phenylthio)butanoic acid
- SMILES: O=C(C[C@H](CSC1=CC=CC=C1)NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.