Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-N-Methyl-1-phenylethylamine-d13, TCI America™
SDP

Catalog No. M17711ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17711ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M17711ML Supplier TCI America Supplier No. M17711ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 357407-86-4
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00463429
InChI Key RCSSHZGQHHEHPZ-UHFFFAOYNA-N
Synonym (R)-(+)-alpha,N-Dimethylbenzylamine-d13
PubChem CID 91659078
IUPAC Name methyl(1-phenylethyl)amine
SMILES CNC(C)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 1 mL
UN Number 2735
Formula Weight 148.29
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material (R)-(+)-N-Methyl-1-phenylethylamine-d13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.