Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-N,N-Dimethyl-1-phenethylamine, ≥99%, Thermo Scientific™

Catalog No. AC221560010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC221560010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC221560010 Supplier Thermo Scientific Chemicals Supplier No. 221560010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19342-01-9
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
InChI Key BVURNMLGDQYNAF-SECBINFHSA-N
Synonym r-+-n,n-dimethyl-1-phenylethylamine, r-n,n-dimethyl-1-phenylethanamine, unii-13x1wu55oq, 1r-n,n-dimethyl-1-phenylethanamine, r-+-n,n-dimethyl-1-phenylethanamine, dimethyl 1r-1-phenylethyl amine, n,n-dimethyl-1-phenylethanamine #, r-+-n,n-dimethyl-1-phenethylamine
PubChem CID 103034
IUPAC Name (1R)-N,N-dimethyl-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N(C)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Density 0.908g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Refractive Index 1.5020 to 1.5040 (20°C, 589nm)
Linear Formula C6H5CH(CH3)N(CH3)2
Quantity 1 g
Beilstein 12, III, 2392
Specific Rotation Condition +50° (20°C c=1,CH3OH)
Specific Rotation +49° to +51° (20°C, 589nm) (c=1, CH3OH)
Formula Weight 149.24
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material (R)-(+)-N, N-Dimethyl-1-phenethylamine, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.