Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-nicardipine-d3 | 00-00-0 | MFCD34676726 | 99.0% | 482.54 g/mol | C26H26D3N3O6 | 5 MG
SDP

Catalog No. 5000264539
Encompass_Preferred

(R)-Nicardipine-d3 is a deuterium-labeled form of R-nicardipine supplied as an analytical reference standard for research use. It is intended for method development, quantitative analysis, and mass spectrometry applications where an isotopically labeled internal standard is required.

  • Deuterium-labeled analog for use as an internal standard in LC-MS assays.
  • High reported purity (99.0%) suitable for analytical applications.
  • Molecular weight 482.54 g/mol and formula C26H26D3N3O6.
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
  • Supplied with a certificate of analysis supporting batch purity and identity.

Catalog No. 50-002-64539 Supplier Medchemexpress LLC Supplier No. HY12515CS15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Nicardipine-d3 is a deuterium-labeled form of R-nicardipine supplied as an analytical reference standard for research use. It is intended for method development, quantitative analysis, and mass spectrometry applications where an isotopically labeled internal standard is required.

  • Deuterium-labeled analog for use as an internal standard in LC-MS assays.
  • High reported purity (99.0%) suitable for analytical applications.
  • Molecular weight 482.54 g/mol and formula C26H26D3N3O6.
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
  • Supplied with a certificate of analysis supporting batch purity and identity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.