Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-Nimodipine | 77940-92-2 | 418.44 | C21H26N2O7 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYB0265A10MG

Encompass_Preferred

(R)-Nimodipine is the R-enantiomer of nimodipine, a dihydropyridine calcium-channel antagonist used in research on cerebrovascular disorders and neuronal signaling. It is supplied as a research-grade small-molecule reagent for pharmacological and mechanistic studies.

  • Chemical formula C21H26N2O7.
  • Molecular weight 418.44.
  • CAS number 77940-92-2.
  • Targets calcium channels and modulates neuronal signaling.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-98886


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.