Learn More
Medchemexpress LLC (R)-Nimodipine | 77940-92-2 | 99.87% | 100 MG
SDP
Catalog No. 5000406188
Encompass_Preferred
(R)-Nimodipine is an orally active, blood-brain barrier-permeable L-type calcium channel blocker with an IC50 of 5 nM. It inhibits corticosterone release by blocking calcium channels on the hypothalamic-pituitary-adrenal axis, which can reverse immobilization stress-induced memory impairment and behavioral abnormalities. It is for research use only.
  • Inhibits corticosterone release
  • Reverses immobilization stress-induced memory impairment
  • Used in studies related to aneurysmal subarachnoid hemorrhage
  • Used in studies related to cerebral ischemia
  • Used in studies related to epilepsy
  • Used in studies related to age-related degenerative neurological diseases
  • Used in studies related to alcohol intoxication
Catalog No. 50-004-06188 Supplier Medchemexpress LLC Supplier No. HYB0265A100MG
(R)-Nimodipine is an orally active, blood-brain barrier-permeable L-type calcium channel blocker with an IC50 of 5 nM. It inhibits corticosterone release by blocking calcium channels on the hypothalamic-pituitary-adrenal axis, which can reverse immobilization stress-induced memory impairment and behavioral abnormalities. It is for research use only.
  • Inhibits corticosterone release
  • Reverses immobilization stress-induced memory impairment
  • Used in studies related to aneurysmal subarachnoid hemorrhage
  • Used in studies related to cerebral ischemia
  • Used in studies related to epilepsy
  • Used in studies related to age-related degenerative neurological diseases
  • Used in studies related to alcohol intoxication