Learn More
Medchemexpress LLC (R)-Nimodipine | 77940-92-2 | 99.9% | 25 MG
SDP
Catalog No. 5000451034
Encompass_Preferred
(R)-Nimodipine is an orally active, blood-brain barrier-permeable L-type calcium channel blocker with an IC50 of 5 nM. It inhibits corticosterone release by blocking calcium channels on the hypothalamic-pituitary-adrenal axis, reversing immobilization stress-induced memory impairment and behavioral abnormalities. This compound is widely used in studies related to aneurysmal subarachnoid hemorrhage, cerebral ischemia, epilepsy, age-related degenerative neurological diseases, and alcohol intoxication.
  • Orally active and blood-brain barrier-permeable
  • L-type calcium channel blocker
  • Inhibits corticosterone release
  • Reverses immobilization stress-induced memory impairment
  • Used in research for neurological and cerebrovascular diseases
  • Inhibits slow inactivating Ca2+ currents in vitro
  • Soluble in DMSO for experimental use
  • Provided with in vivo protocols for various formulations
Catalog No. 50-004-51034 Supplier Medchemexpress LLC Supplier No. 25MGHYB0265A
(R)-Nimodipine is an orally active, blood-brain barrier-permeable L-type calcium channel blocker with an IC50 of 5 nM. It inhibits corticosterone release by blocking calcium channels on the hypothalamic-pituitary-adrenal axis, reversing immobilization stress-induced memory impairment and behavioral abnormalities. This compound is widely used in studies related to aneurysmal subarachnoid hemorrhage, cerebral ischemia, epilepsy, age-related degenerative neurological diseases, and alcohol intoxication.
  • Orally active and blood-brain barrier-permeable
  • L-type calcium channel blocker
  • Inhibits corticosterone release
  • Reverses immobilization stress-induced memory impairment
  • Used in research for neurological and cerebrovascular diseases
  • Inhibits slow inactivating Ca2+ currents in vitro
  • Soluble in DMSO for experimental use
  • Provided with in vivo protocols for various formulations