Learn More
Medchemexpress LLC HY-114395A 10mg Medchemexpress, (R)-NVS-ZP7-4 CAS: Purity:>98%
SDP
Catalog No. 501969343
Encompass_Preferred
Medchemexpress, HY-114395A 10mg (R)-NVS-ZP7-4 CAS: (R)-NVS-ZP7-4 is the R-isomer of NVS-ZP7-4. NVS-ZP7-4 is a Zinc transporter SLC39A7 (ZIP7) inhibitor that is also the first reported chemical tool to probe the impact of modulating ER zinc levels and investigate ZIP7 as a novel druggable node in the Notch pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9343 Supplier Medchemexpress LLC Supplier No. HY114395A10MG
Medchemexpress, HY-114395A 10mg (R)-NVS-ZP7-4 CAS: (R)-NVS-ZP7-4 is the R-isomer of NVS-ZP7-4. NVS-ZP7-4 is a Zinc transporter SLC39A7 (ZIP7) inhibitor that is also the first reported chemical tool to probe the impact of modulating ER zinc levels and investigate ZIP7 as a novel druggable node in the Notch pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.