Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-NX-2127 | 3024312-52-2 | 98.9% | 719.83 | C39H45N9O5 | 10 MG
SDP

Catalog No. 5000286015
Encompass_Preferred

(R)-NX-2127 is the R-isomer of a Bruton's tyrosine kinase (BTK) PROTAC degrader supplied as a high-purity solid for preclinical research. The compound is intended for use in studies of BTK degradation and related cellular pharmacology; consult the supplier product page and SDS for handling and storage details.

  • R-isomer of a BTK PROTAC degrader
  • High reported purity (98.93%)
  • Molecular weight 719.83
  • Chemical formula C39H45N9O5
  • Solid, light yellow to yellow appearance
  • Recommended storage: 4°C protected from light; in solvent -80°C (6 months) or -20°C (1 month)
  • Intended for preclinical BTK degradation and cellular assays

Catalog No. 50-002-86015 Supplier Medchemexpress LLC Supplier No. HY153220A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-NX-2127 is the R-isomer of a Bruton's tyrosine kinase (BTK) PROTAC degrader supplied as a high-purity solid for preclinical research. The compound is intended for use in studies of BTK degradation and related cellular pharmacology; consult the supplier product page and SDS for handling and storage details.

  • R-isomer of a BTK PROTAC degrader
  • High reported purity (98.93%)
  • Molecular weight 719.83
  • Chemical formula C39H45N9O5
  • Solid, light yellow to yellow appearance
  • Recommended storage: 4°C protected from light; in solvent -80°C (6 months) or -20°C (1 month)
  • Intended for preclinical BTK degradation and cellular assays

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.