Learn More
Cayman Chemical (R)-Ofloxacin, Purity: ≥98%, CAS: 100986-86-5, 5 mg
SDP
Catalog No. 502989559
Encompass_Preferred
A fluoroquinolone antibiotic; the (R) isomer of ofloxacin and levofloxacin; active against certain Gram-positive and Gram-negative bacteria, including E. coli, P. aeruginosa strains 32104 and 32122, S. aureus strains 209P and Smith, and S. epidermis strain 56556 (MICs = 0.78, 12.5, 6.25, 25, 12.5, and 25 µg/ml, respectively) but not S. epidermis strain 56500, S. pyogenes, or S. faecalis (MICs = >100 µg/ml for all); inhibits E. coli DNA gyrase with an IC50 value of 75 µg/ml, which is approximately 30- and 50-fold lower than inhibition by ofloxacin and levofloxacin, respectively
Catalog No. 50-298-9559 Supplier Cayman Chemical Supplier No. 296015 MG
A fluoroquinolone antibiotic; the (R) isomer of ofloxacin and levofloxacin; active against certain Gram-positive and Gram-negative bacteria, including E. coli, P. aeruginosa strains 32104 and 32122, S. aureus strains 209P and Smith, and S. epidermis strain 56556 (MICs = 0.78, 12.5, 6.25, 25, 12.5, and 25 µg/ml, respectively) but not S. epidermis strain 56500, S. pyogenes, or S. faecalis (MICs = >100 µg/ml for all); inhibits E. coli DNA gyrase with an IC50 value of 75 µg/ml, which is approximately 30- and 50-fold lower than inhibition by ofloxacin and levofloxacin, respectively