Learn More
Medchemexpress LLC (R)-Omeprazole sodium | 161796-77-6 | 98.1% | 100 MG
SDP
Catalog No. 5000338526
Encompass_Preferred
(R)-Omeprazole sodium is a gastric acid resistant compound that inhibits gastric acid secretion. It is primarily metabolized by cytochrome P450 enzymes in vivo, and its interaction with mannitol may affect its bioavailability in formulations. This compound exhibits reversible direct and metabolism-dependent inhibition of CYP2C19. It is intended for research use only.
  • Gastric acid resistant compound
  • Inhibits gastric acid secretion
  • Metabolism affected by cytochrome P450 enzymes
  • May interact with mannitol affecting bioavailability
  • Exhibits reversible direct and metabolism-dependent inhibition of CYP2C19
  • For research use only
Catalog No. 50-003-38526 Supplier Medchemexpress LLC Supplier No. HY129923100MG
(R)-Omeprazole sodium is a gastric acid resistant compound that inhibits gastric acid secretion. It is primarily metabolized by cytochrome P450 enzymes in vivo, and its interaction with mannitol may affect its bioavailability in formulations. This compound exhibits reversible direct and metabolism-dependent inhibition of CYP2C19. It is intended for research use only.
  • Gastric acid resistant compound
  • Inhibits gastric acid secretion
  • Metabolism affected by cytochrome P450 enzymes
  • May interact with mannitol affecting bioavailability
  • Exhibits reversible direct and metabolism-dependent inhibition of CYP2C19
  • For research use only