Learn More
Medchemexpress LLC (R)-Omeprazole (sodium) | 161796-77-6 | 98.1% | 25 MG
SDP
Catalog No. 5000342213
Encompass_Preferred
(R)-Omeprazole sodium is a gastric acid resistant compound that inhibits gastric acid secretion. It is metabolized in vivo, primarily by cytochrome P450 enzymes, and its interaction with mannitol may affect its bioavailability in formulations. It exhibits reversible direct and metabolism-dependent inhibition of CYP2C19.
  • Gastric acid resistant compound
  • Inhibits gastric acid secretion
  • Metabolized by cytochrome P450 enzymes
  • Reversible direct and metabolism-dependent inhibition of CYP2C19
  • For research use only
Catalog No. 50-003-42213 Supplier Medchemexpress LLC Supplier No. 25MGHY129923
(R)-Omeprazole sodium is a gastric acid resistant compound that inhibits gastric acid secretion. It is metabolized in vivo, primarily by cytochrome P450 enzymes, and its interaction with mannitol may affect its bioavailability in formulations. It exhibits reversible direct and metabolism-dependent inhibition of CYP2C19.
  • Gastric acid resistant compound
  • Inhibits gastric acid secretion
  • Metabolized by cytochrome P450 enzymes
  • Reversible direct and metabolism-dependent inhibition of CYP2C19
  • For research use only