Learn More
Medchemexpress LLC (R)-Omeprazole (sodium) | 161796-77-6 | 98.1% | 250 MG
SDP
Catalog No. 5000341984
Encompass_Preferred
(R)-Omeprazole sodium is a gastric acid resistant compound that inhibits gastric acid secretion. It is primarily metabolized in vivo by cytochrome P450 enzymes. Interactions with mannitol may affect its bioavailability in certain formulations.
  • Inhibits gastric acid secretion
  • Metabolized by cytochrome P450 enzymes
  • Interacts with mannitol
  • Reversibly inhibits CYP2C19 directly and metabolism-dependently
  • For research use only
Catalog No. 50-003-41984 Supplier Medchemexpress LLC Supplier No. 250MGHY129923
(R)-Omeprazole sodium is a gastric acid resistant compound that inhibits gastric acid secretion. It is primarily metabolized in vivo by cytochrome P450 enzymes. Interactions with mannitol may affect its bioavailability in certain formulations.
  • Inhibits gastric acid secretion
  • Metabolized by cytochrome P450 enzymes
  • Interacts with mannitol
  • Reversibly inhibits CYP2C19 directly and metabolism-dependently
  • For research use only