Learn More
Medchemexpress LLC (R)-Omeprazole sodium | 161796-77-6 | 98.1% | 50 MG
SDP
Catalog No. 5000338527
Encompass_Preferred
(R)-Omeprazole sodium is a gastric acid resistant compound that inhibits gastric acid secretion. Its metabolism in vivo is primarily affected by cytochrome P450 enzymes. The interaction between (R)-Omeprazole sodium and mannitol may affect its bioavailability in formulations. It exhibits reversible direct and metabolism-dependent inhibition of CYP2C19.
  • Gastric acid resistant compound
  • Inhibits gastric acid secretion
  • Metabolized primarily by cytochrome P450 enzymes
  • Interaction with mannitol may affect bioavailability
  • Exhibits reversible direct and metabolism-dependent inhibition of CYP2C19
Catalog No. 50-003-38527 Supplier Medchemexpress LLC Supplier No. HY12992350MG
(R)-Omeprazole sodium is a gastric acid resistant compound that inhibits gastric acid secretion. Its metabolism in vivo is primarily affected by cytochrome P450 enzymes. The interaction between (R)-Omeprazole sodium and mannitol may affect its bioavailability in formulations. It exhibits reversible direct and metabolism-dependent inhibition of CYP2C19.
  • Gastric acid resistant compound
  • Inhibits gastric acid secretion
  • Metabolized primarily by cytochrome P450 enzymes
  • Interaction with mannitol may affect bioavailability
  • Exhibits reversible direct and metabolism-dependent inhibition of CYP2C19