Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-omeprazole sodium | 161796-77-6 | 98.1% | C17H18N3NaO3S | 10MG
SDP

Catalog No. 5000260902
Encompass_Preferred

(R)-omeprazole sodium is the R-enantiomer of omeprazole supplied as the sodium salt. It is a gastric acid-resistant proton pump inhibitor used as a research reference standard to study gastric acid secretion and CYP2C19-mediated metabolism. The compound is provided with high chemical and enantiomeric purity for analytical and pharmacology studies.

  • Molecular formula: C17H18N3NaO3S.
  • Molecular weight: 367.4 g/mol.
  • Purity: 98.1% (reported 98.07%).
  • Enantiomeric excess: 99.84%.
  • Function: reversible and metabolism-dependent inhibitor of CYP2C19.
  • Intended use: reference standard and research compound for pharmacology and metabolism studies.
  • Availability: includes small-scale 10 mg quantities suitable for analytical work.

Catalog No. 50-002-60902 Supplier Medchemexpress LLC Supplier No. HY12992310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-omeprazole sodium is the R-enantiomer of omeprazole supplied as the sodium salt. It is a gastric acid-resistant proton pump inhibitor used as a research reference standard to study gastric acid secretion and CYP2C19-mediated metabolism. The compound is provided with high chemical and enantiomeric purity for analytical and pharmacology studies.

  • Molecular formula: C17H18N3NaO3S.
  • Molecular weight: 367.4 g/mol.
  • Purity: 98.1% (reported 98.07%).
  • Enantiomeric excess: 99.84%.
  • Function: reversible and metabolism-dependent inhibitor of CYP2C19.
  • Intended use: reference standard and research compound for pharmacology and metabolism studies.
  • Availability: includes small-scale 10 mg quantities suitable for analytical work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.