Learn More
Medchemexpress LLC (R)-ONO-2952 | 2988224-39-9 | MFCD34676563 | 99.8% | 398.86 | C22H20ClFN2O2 | 5 MG
SDP
Catalog No. 5000264313
Encompass_Preferred
(R)-ONO-2952 is the R-enantiomer of ONO-2952, a translocator protein 18 kDa (TSPO) antagonist developed for research into stress-related and irritable bowel syndrome indications. Supplied as a white to off-white solid, the compound has molecular formula C22H20ClFN2O2, molecular weight 398.86, and supplier-reported purity of approximately 99.8%, with recommended low-temperature storage for long-term stability.
  • High reported purity suitable for analytical and pharmacology studies.
  • R-enantiomer provided for stereochemistry-specific experiments.
  • Solid powder form with defined storage conditions for stability.
  • Documented molecular formula and molecular weight for formulation and dosing calculations.
  • Suitable as a reference standard in preclinical TSPO research.
Catalog No. 50-002-64313 Supplier Medchemexpress LLC Supplier No. HY111191A5MG
(R)-ONO-2952 is the R-enantiomer of ONO-2952, a translocator protein 18 kDa (TSPO) antagonist developed for research into stress-related and irritable bowel syndrome indications. Supplied as a white to off-white solid, the compound has molecular formula C22H20ClFN2O2, molecular weight 398.86, and supplier-reported purity of approximately 99.8%, with recommended low-temperature storage for long-term stability.
  • High reported purity suitable for analytical and pharmacology studies.
  • R-enantiomer provided for stereochemistry-specific experiments.
  • Solid powder form with defined storage conditions for stability.
  • Documented molecular formula and molecular weight for formulation and dosing calculations.
  • Suitable as a reference standard in preclinical TSPO research.