Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(p-Toluenesulfinyl)ferrocene 98.0+%, TCI America™
SDP

Catalog No. T25455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25455G 5 g
T25451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25455G Supplier TCI America Supplier No. T25455G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 130225-27-3
Molecular Formula C17H16FeOS
Synonym (R)-Ferrocenyl p-Tolyl Sulfoxide

Specifications

Melting Point 143°C
Quantity 5 g
Formula Weight 324.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(p-Toluenesulfinyl)ferrocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.