Learn More
Medchemexpress LLC (R)-palmitoyl-(2-methyl)ethanolamide | 179951-56-5 | MFCD02259379 | 99.0% | 313.52 g·mol⁻¹ | C19H39NO2 | 10 MG
SDP
Catalog No. 5000288351
Encompass_Preferred
(R)-Palmitoyl-(2-methyl)ethanolamide (Me-PEA) is the R-enantiomer of palmitoyl-(2-methyl)ethanolamide used as a biochemical research reagent. It acts as a competitive inhibitor of [3H]-anandamide (AEA) metabolism and is employed to probe endocannabinoid metabolic pathways.
  • Competitive inhibitor of [3H]-AEA metabolism; reported Ki = 6.6 μM.
  • High purity (99.0%) suitable for biochemical assays.
  • Molecular formula C19H39NO2 and molecular weight 313.52 g·mol⁻¹.
  • Solid appearance; stable as powder at -20°C for up to 3 years.
  • Intended for research use to study endocannabinoid metabolism and related enzymatic pathways.
Catalog No. 50-002-88351 Supplier Medchemexpress LLC Supplier No. HY15782910MG
(R)-Palmitoyl-(2-methyl)ethanolamide (Me-PEA) is the R-enantiomer of palmitoyl-(2-methyl)ethanolamide used as a biochemical research reagent. It acts as a competitive inhibitor of [3H]-anandamide (AEA) metabolism and is employed to probe endocannabinoid metabolic pathways.
  • Competitive inhibitor of [3H]-AEA metabolism; reported Ki = 6.6 μM.
  • High purity (99.0%) suitable for biochemical assays.
  • Molecular formula C19H39NO2 and molecular weight 313.52 g·mol⁻¹.
  • Solid appearance; stable as powder at -20°C for up to 3 years.
  • Intended for research use to study endocannabinoid metabolism and related enzymatic pathways.