Learn More
Medchemexpress LLC (R)-pantetheine | 496-65-1 | 99.0% | 278.37 g·mol⁻¹ | C11H22N2O4S | 5MG
SDP
Catalog No. 5000239131
Encompass_Preferred
(R)-Pantetheine is the R-enantiomer of pantetheine and the direct biosynthetic precursor to coenzyme A (CoA). It is supplied as a high-purity research reagent and analytical standard for studies of metabolism and CoA biosynthesis. Supplied in milligram-scale quantities for laboratory use; not for human consumption and may be toxic.
  • Used for studies of CoA biosynthesis and cellular metabolism.
  • Supplied as an analytical standard for quantitative and qualitative assays.
  • High purity (99.0%).
  • Known molecular formula C11H22N2O4S and molecular weight 278.37 g·mol⁻¹.
  • Available in small milligram-scale pack sizes suitable for analytical workflows.
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-39131 Supplier Medchemexpress LLC Supplier No. HY1260505MG
(R)-Pantetheine is the R-enantiomer of pantetheine and the direct biosynthetic precursor to coenzyme A (CoA). It is supplied as a high-purity research reagent and analytical standard for studies of metabolism and CoA biosynthesis. Supplied in milligram-scale quantities for laboratory use; not for human consumption and may be toxic.
  • Used for studies of CoA biosynthesis and cellular metabolism.
  • Supplied as an analytical standard for quantitative and qualitative assays.
  • High purity (99.0%).
  • Known molecular formula C11H22N2O4S and molecular weight 278.37 g·mol⁻¹.
  • Available in small milligram-scale pack sizes suitable for analytical workflows.
  • For research use only; not for human or clinical applications.