Learn More
Med Vet International Miltex Vantage Rochester-Pean Forceps, 6.25in, Straight
Supplier:  Med Vet International MILV97120
Encompass_Preferred
The Miltex Vantage V97-120 Rochester-Pean Forceps have a length of 6.25in. and are made of mid-grade stainless steel. They have a straight tip and are a hemostatic forcep with fully serrated jaws. This surgical forcep is commonly used to occlude or clamp blood vessels to control bleeding during surgery.
Catalog No. 50-255-9439