Learn More
Cayman Chemical R-PFI-2hydrochlorIde 10mg
SDP
Catalog No. 502968548
Encompass_Preferred
A potent, cell-permeable inhibitor of SET7/9 (IC50 = 2 nM) that demonstrates greater than 1,000-fold selectivity over a panel of 18 other methyltransferases
Catalog No. 50-296-8548 Supplier Cayman Chemical Supplier No. 1467810 MG
A potent, cell-permeable inhibitor of SET7/9 (IC50 = 2 nM) that demonstrates greater than 1,000-fold selectivity over a panel of 18 other methyltransferases