Learn More
Cayman Chemical (R)-PFI-2 (hydrochloride), Purity: ≥95%, CAS: 1627607-87-7, 5 mg
SDP
Catalog No. 502969486
Encompass_Preferred
A potent, cell-permeable inhibitor of SET7/9 (IC50 = 2 nM) that demonstrates greater than 1,000-fold selectivity over a panel of 18 other methyltransferases
Catalog No. 50-296-9486 Supplier Cayman Chemical Supplier No. 146785 MG
A potent, cell-permeable inhibitor of SET7/9 (IC50 = 2 nM) that demonstrates greater than 1,000-fold selectivity over a panel of 18 other methyltransferases