Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-Phanephos, 98%, ee 98%, Thermo Scientific™

Catalog No. AAH25812MD
Click to view available options
Quantity:
250mg

Catalog No. AAH25812MD

Supplier: Thermo Scientific Chemicals H25812MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 364732-88-7
Molecular Formula C40H34P2
Quantity 250mg
MDL Number MFCD03094575
Synonym r-phanephos, phanephos, s-phanephos, 2.2 phanephos, r---4,12-bis diphenylphosphino-2.2-paracyclophane, s-+-4,12-bis diphenylphosphino-2.2-paracyclophane, phanepho s, bisdiphenylphosphinoparacyclophane, s-+-4,12-bis-diphenylphosphino-2.2-paracyclophane
InChI Key GYZZZILPVUYAFJ-UHFFFAOYSA-N
SMILES C1CC2=C(C=C(CCC3=C(C=C1C=C3)P(C4=CC=CC=C4)C5=CC=CC=C5)C=C2)P(C6=CC=CC=C6)C7=CC=CC=C7
Molecular Weight (g/mol) 576.66
PubChem CID 10864772
Formula Weight 576.65
Percent Purity 98%
Assay ee 98%
Chemical Name or Material (R)-Phanephos
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.