Learn More
Medchemexpress LLC (R)-(-)-α-Phellandrene | 4221-98-1 | 85.8% | 136.23 | 1 ML
SDP
Catalog No. 5000426417
Encompass_Preferred
(R)-(-)-α-Phellandrene, also known as (-)-α-Phellandrene, is an orally active cyclic monoterpene and the (R)-(-)-stereoisomer of α-phellandrene. It has demonstrated the ability to attenuate inflammatory responses and induce DNA damage. This product is for research use only.
  • Attenuates inflammatory responses
  • Induces DNA damage
  • Shows weak inhibitory activities against bacteria (e.g., MRSA, A. tumefaciens, and E. carotovora subsp. Carotovora)
  • Colorless to light yellow liquid with a density of 0.844 g/cm³
  • Classified as a terpenoid, specifically an other monoterpene
  • Initial source is plants, particularly Anacardiaceae Schinus terebinthifolius Raddi
  • Ships at room temperature in continental US; stores at 4°C under nitrogen
Catalog No. 50-004-26417 Supplier Medchemexpress LLC Supplier No. HYN94871ML
(R)-(-)-α-Phellandrene, also known as (-)-α-Phellandrene, is an orally active cyclic monoterpene and the (R)-(-)-stereoisomer of α-phellandrene. It has demonstrated the ability to attenuate inflammatory responses and induce DNA damage. This product is for research use only.
  • Attenuates inflammatory responses
  • Induces DNA damage
  • Shows weak inhibitory activities against bacteria (e.g., MRSA, A. tumefaciens, and E. carotovora subsp. Carotovora)
  • Colorless to light yellow liquid with a density of 0.844 g/cm³
  • Classified as a terpenoid, specifically an other monoterpene
  • Initial source is plants, particularly Anacardiaceae Schinus terebinthifolius Raddi
  • Ships at room temperature in continental US; stores at 4°C under nitrogen