Learn More
Medchemexpress LLC (R)-phenyl (5-(2-(2,5-difluorophenyl)pyrrolidin-1-yl)pyrazolo[1,5-a]pyrimidin-3-yl)carbamate | 2135871-21-3 | 99.5% | 435.43 g·mol⁻1 | C23H19F2N5O2 | 10 MG
SDP
Catalog No. 5000299160
Encompass_Preferred
(R)-Phenyl (5-(2-(2,5-difluorophenyl)pyrrolidin-1-yl)pyrazolo[1,5-a]pyrimidin-3-yl)carbamate is an (R)-enantiomeric chemical intermediate for use in medicinal chemistry and synthetic workflows. It is intended for research-scale synthesis of pyrazolo[1,5-a]pyrimidine-derived active compounds and is supplied with analytical documentation for characterization.
  • High analytical purity (99.5%).
  • Supplied as the (R)-enantiomer for enantiospecific synthesis.
  • Intended for use as a drug intermediate in medicinal chemistry.
  • Available in research-scale pack sizes, including 10 MG and sample quantities.
  • Storage stability: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Analytical documentation available, including COA and SDS.
Catalog No. 50-002-99160 Supplier Medchemexpress LLC Supplier No. HY4269710MG
(R)-Phenyl (5-(2-(2,5-difluorophenyl)pyrrolidin-1-yl)pyrazolo[1,5-a]pyrimidin-3-yl)carbamate is an (R)-enantiomeric chemical intermediate for use in medicinal chemistry and synthetic workflows. It is intended for research-scale synthesis of pyrazolo[1,5-a]pyrimidine-derived active compounds and is supplied with analytical documentation for characterization.
  • High analytical purity (99.5%).
  • Supplied as the (R)-enantiomer for enantiospecific synthesis.
  • Intended for use as a drug intermediate in medicinal chemistry.
  • Available in research-scale pack sizes, including 10 MG and sample quantities.
  • Storage stability: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Analytical documentation available, including COA and SDS.